Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNAP47
Entrez Gene ID
101104060
Gene Name
synaptosome associated protein 47
Gene Aliases
-
Chromosome Location
Chr.5: 3416424 - 3484643 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004008398.4 XP_004008447.3 1 - 2 227 74
XM_012177653.3 XP_012033043.3 1 - 2 239 74

View Full Product Details