Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSPB8
Entrez Gene ID
101108164
Gene Name
heat shock protein family B (small) member 8
Gene Aliases
-
Chromosome Location
Chr.17: 65105441 - 65118554 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017375.4 XP_004017424.2 2 - 3 1001 70

View Full Product Details