Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHM
Entrez Gene ID
101102290
Gene Name
CHM Rab escort protein
Gene Aliases
-
Chromosome Location
Chr.X: 84045216 - 84294715 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004022232.4 XP_004022281.1 9 - 10 1266 109

View Full Product Details