Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HNRNPR
Entrez Gene ID
101121577
Gene Name
heterogeneous nuclear ribonucleoprotein R
Gene Aliases
-
Chromosome Location
Chr.2: 259033246 - 259066856 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004005133.4 XP_004005182.2 7 - 8 914 114
XM_027965541.1 XP_027821342.1 7 - 8 910 114
XM_027965542.1 XP_027821343.1 6 - 7 766 114

Gene Symbol
LOC114118352
Entrez Gene ID
114118352
Gene Name
heterogeneous nuclear ribonucleoprotein R-like
Gene Aliases
-
Chromosome Location
Chr.15: 165975 - 222332 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027979098.1 XP_027834899.1 5 - 5 972 114

View Full Product Details