Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GUCA1B
Entrez Gene ID
101108609
Gene Name
guanylate cyclase activator 1B
Gene Aliases
-
Chromosome Location
Chr.20: 17655070 - 17665695 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004018819.4 XP_004018868.2 2 - 3 357 75

View Full Product Details