Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LYZL6
Entrez Gene ID
101119574
Gene Name
lysozyme like 6
Gene Aliases
-
Chromosome Location
Chr.11: 17840871 - 17845267 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975440.1 XP_027831241.1 2 - 3 300 72

View Full Product Details