Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP4R2
Entrez Gene ID
101113890
Gene Name
protein phosphatase 4 regulatory subunit 2
Gene Aliases
-
Chromosome Location
Chr.19: 29970731 - 30023979 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015102332.2 XP_014957818.1 2 - 3 338 121

View Full Product Details