Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DEAF1
Entrez Gene ID
101103106
Gene Name
DEAF1 transcription factor
Gene Aliases
-
Chromosome Location
Chr.21: 52355105 - 52379873 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959954.1 XP_027815755.1 9 - 10 1285 112

View Full Product Details