Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPP1R3A
Entrez Gene ID
101110321
Gene Name
protein phosphatase 1 regulatory subunit 3A
Gene Aliases
-
Chromosome Location
Chr.4: 61091473 - 61134183 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015095296.2 XP_014950782.2 1 - 2 813 119

View Full Product Details