Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
METTL11B
Entrez Gene ID
101103252
Gene Name
methyltransferase like 11B
Gene Aliases
-
Chromosome Location
Chr.12: 41074977 - 41093781 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013682.4 XP_004013731.2 1 - 2 270 91

Gene Symbol
LOC114117224
Entrez Gene ID
114117224
Gene Name
alpha N-terminal protein methyltransferase 1B-like
Gene Aliases
-
Chromosome Location
Chr.12: 43575416 - 43599362 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975622.1 XP_027831423.1 1 - 2 270 91

View Full Product Details