Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AIPL1
Entrez Gene ID
101104920
Gene Name
aryl hydrocarbon receptor interacting protein like 1
Gene Aliases
-
Chromosome Location
Chr.11: 37925642 - 37935249 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012585.4 XP_004012634.2 3 - 4 495 101

View Full Product Details