Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
XKR4
Entrez Gene ID
101108498
Gene Name
XK related 4
Gene Aliases
-
Chromosome Location
Chr.9: 38381839 - 38703650 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027972963.1 XP_027828764.1 1 - 2 835 76

View Full Product Details