Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SELL
Entrez Gene ID
101102077
Gene Name
selectin L
Gene Aliases
-
Chromosome Location
Chr.12: 40644155 - 40669607 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013677.4 XP_004013726.1 4 - 5 835 72

View Full Product Details