Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPM1E
Entrez Gene ID
101106345
Gene Name
protein phosphatase, Mg2+/Mn2+ dependent 1E
Gene Aliases
-
Chromosome Location
Chr.11: 53997213 - 54221938 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975349.1 XP_027831150.1 4 - 5 978 100

View Full Product Details