Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
VPS28
Entrez Gene ID
101107800
Gene Name
VPS28 subunit of ESCRT-I
Gene Aliases
-
Chromosome Location
Chr.9: 14722832 - 14727270 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027973435.1 XP_027829236.1 9 - 10 821 108
XM_027973436.1 XP_027829237.1 9 - 10 813 108
XM_027973437.1 XP_027829238.1 8 - 9 588 108
XM_027973438.1 XP_027829239.1 8 - 9 688 108
XM_027973439.1 XP_027829240.1 8 - 9 687 108

View Full Product Details