Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MAB21L3
Entrez Gene ID
101110381
Gene Name
mab-21 like 3
Gene Aliases
-
Chromosome Location
Chr.1: 98276799 - 98305754 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027973497.1 XP_027829298.1 5 - 6 666 66
XM_027973498.1 XP_027829299.1 5 - 6 682 66
XM_027973499.1 XP_027829300.1 2 - 3 429 66

View Full Product Details