Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDF2
Entrez Gene ID
101106183
Gene Name
stromal cell derived factor 2
Gene Aliases
-
Chromosome Location
Chr.11: 43275840 - 43283872 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012502.2 XP_004012551.1 1 - 2 428 93

View Full Product Details