Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB25
Entrez Gene ID
101123339
Gene Name
RAB25, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.1: 112246504 - 112253333 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976329.1 XP_027832130.1 4 - 5 769 58

View Full Product Details