Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYLC2
Entrez Gene ID
101120048
Gene Name
cylicin 2
Gene Aliases
-
Chromosome Location
Chr.2: 20874808 - 21047510 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964011.1 XP_027819812.1 3 - 4 229 82

View Full Product Details