Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FICD
Entrez Gene ID
101103594
Gene Name
FIC domain protein adenylyltransferase
Gene Aliases
-
Chromosome Location
Chr.17: 73633068 - 73638281 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012097958.2 XP_011953348.1 2 - 3 447 66

View Full Product Details