Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC101113346
Entrez Gene ID
101113346
Gene Name
schlafen family member 13
Gene Aliases
SLFN11, SLFN12L, SLFN13
Chromosome Location
Chr.11: 48899392 - 48963934 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975299.1 XP_027831100.1 1 - 2 240 69

View Full Product Details