Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FBXO45
Entrez Gene ID
101123179
Gene Name
F-box protein 45
Gene Aliases
-
Chromosome Location
Chr.1: 209144134 - 209159007 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015092481.2 XP_014947967.2 1 - 2 564 81

View Full Product Details