Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
VPS37A
Entrez Gene ID
101120049
Gene Name
VPS37A subunit of ESCRT-I
Gene Aliases
-
Chromosome Location
Chr.26: 21482607 - 21519762 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004021731.4 XP_004021780.1 2 - 3 538 94

View Full Product Details