Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
POLD4
Entrez Gene ID
101118743
Gene Name
DNA polymerase delta 4, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.21: 47011192 - 47012980 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004019714.4 XP_004019763.1 1 - 2 304 115

View Full Product Details