Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RPA3
Entrez Gene ID
101121087
Gene Name
replication protein A3
Gene Aliases
-
Chromosome Location
Chr.4: 17710349 - 17713636 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027968480.1 XP_027824281.1 2 - 3 228 121

View Full Product Details