Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COMMD6
Entrez Gene ID
101102498
Gene Name
COMM domain containing 6
Gene Aliases
-
Chromosome Location
Chr.10: 54549562 - 54571388 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004012187.4 XP_004012236.2 3 - 4 262 99

View Full Product Details