Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
HSF5
Entrez Gene ID
101105087
Gene Name
heat shock transcription factor 5
Gene Aliases
-
Chromosome Location
Chr.11: 54498424 - 54549563 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004013175.4 XP_004013224.3 3 - 4 1094 60

View Full Product Details