Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGPP2
Entrez Gene ID
101116130
Gene Name
sphingosine-1-phosphate phosphatase 2
Gene Aliases
-
Chromosome Location
Chr.2: 239560887 - 239715924 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093553.2 XP_014949039.2 2 - 3 434 109

View Full Product Details