Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARD6G
Entrez Gene ID
101121649
Gene Name
par-6 family cell polarity regulator gamma
Gene Aliases
-
Chromosome Location
Chr.23: 494243 - 570565 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004020391.4 XP_004020440.4 2 - 3 391 78

View Full Product Details