Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGO2
Entrez Gene ID
101104300
Gene Name
shugoshin 2
Gene Aliases
-
Chromosome Location
Chr.2: 216815204 - 216844864 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012142023.2 XP_011997413.1 6 - 7 821 154

View Full Product Details