Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MPPED1
Entrez Gene ID
101116897
Gene Name
metallophosphoesterase domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 235141247 - 235217881 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015095056.2 XP_014950542.2 1 - 2 613 91

View Full Product Details