Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SPIN1
Entrez Gene ID
101102870
Gene Name
spindlin 1
Gene Aliases
-
Chromosome Location
Chr.2: 24510334 - 24592852 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015093019.2 XP_014948505.1 2 - 3 1232 83
XM_027964174.1 XP_027819975.1 2 - 3 310 83
XM_027964175.1 XP_027819976.1 3 - 4 1647 83

View Full Product Details