Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PCDH18
Entrez Gene ID
101118052
Gene Name
protocadherin 18
Gene Aliases
-
Chromosome Location
Chr.17: 21889722 - 21904503 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004017250.4 XP_004017299.1 1 - 2 2911 90
XM_027956274.1 XP_027812075.1 2 - 3 2656 90

View Full Product Details