Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM14
Entrez Gene ID
101103791
Gene Name
tripartite motif containing 14
Gene Aliases
-
Chromosome Location
Chr.2: 52307087 - 52334671 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015092779.2 XP_014948265.2 4 - 5 743 64

View Full Product Details