Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZNF41
Entrez Gene ID
101116888
Gene Name
zinc finger protein 41
Gene Aliases
-
Chromosome Location
Chr.X: 59673771 - 59715834 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004023243.4 XP_004023292.3 2 - 3 380 64

View Full Product Details