Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CXHXorf58
Entrez Gene ID
101114165
Gene Name
chromosome X CXorf58 homolog
Gene Aliases
-
Chromosome Location
Chr.X: 23580314 - 23603717 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027963191.1 XP_027818992.1 1 - 2 402 87
XM_027963192.1 XP_027818993.1 3 - 4 325 87

View Full Product Details