Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CACNG6
Entrez Gene ID
101118891
Gene Name
calcium voltage-gated channel auxiliary subunit gamma 6
Gene Aliases
-
Chromosome Location
Chr.14: 66217596 - 66233854 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_004015414.3 XP_004015463.2 2 - 3 1598 91

View Full Product Details