Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BTBD19
Entrez Gene ID
101110018
Gene Name
BTB domain containing 19
Gene Aliases
-
Chromosome Location
Chr.1: 20152466 - 20157996 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027969145.1 XP_027824946.1 2 - 3 703 94

View Full Product Details