Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MRPS26
Entrez Gene ID
101112501
Gene Name
mitochondrial ribosomal protein S26
Gene Aliases
-
Chromosome Location
Chr.13: 54232307 - 54235001 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027976870.1 XP_027832671.1 3 - 4 899 68

View Full Product Details