Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KLHDC7B
Entrez Gene ID
101118522
Gene Name
kelch domain containing 7B
Gene Aliases
-
Chromosome Location
Chr.3: 240902848 - 240906315 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027966014.1 XP_027821815.1 1 - 1 2235 86

View Full Product Details