Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCL2L10
Entrez Gene ID
101106530
Gene Name
BCL2 like 10
Gene Aliases
-
Chromosome Location
Chr.7: 60005795 - 60008769 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012181549.3 XP_012036939.3 1 - 2 428 99

View Full Product Details