Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FDX1
Entrez Gene ID
101106360
Gene Name
ferredoxin 1
Gene Aliases
-
Chromosome Location
Chr.15: 22461026 - 22493921 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015100905.2 XP_014956391.2 4 - 5 579 86

View Full Product Details