Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRMT61A
Entrez Gene ID
101102182
Gene Name
tRNA methyltransferase 61A
Gene Aliases
-
Chromosome Location
Chr.18: 68870565 - 68876943 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957446.1 XP_027813247.1 2 - 3 434 81

View Full Product Details