Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MS4A18
Entrez Gene ID
101120872
Gene Name
membrane spanning 4-domains A18
Gene Aliases
-
Chromosome Location
Chr.21: 38135476 - 38148798 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027959682.1 XP_027815483.1 2 - 3 631 111

View Full Product Details