Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TIMP4
Entrez Gene ID
105603615
Gene Name
TIMP metallopeptidase inhibitor 4
Gene Aliases
-
Chromosome Location
Chr.19: 58833862 - 58844888 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957804.1 XP_027813605.1 4 - 5 546 81

View Full Product Details