Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MEGF9
Entrez Gene ID
101116545
Gene Name
multiple EGF like domains 9
Gene Aliases
-
Chromosome Location
Chr.2: 2870977 - 2953795 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964060.1 XP_027819861.1 3 - 4 1115 71

View Full Product Details