Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LOC114113126
Entrez Gene ID
114113126
Gene Name
chondroitin sulfate proteoglycan 5-like
Gene Aliases
-
Chromosome Location
Chr.2: 155145356 - 155158930 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027964809.1 XP_027820610.1 2 - 2 1119 87

Gene Symbol
CSPG5
Entrez Gene ID
101122060
Gene Name
chondroitin sulfate proteoglycan 5
Gene Aliases
-
Chromosome Location
Chr.19: 54044079 - 54055934 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027957898.1 XP_027813699.1 3 - 4 1500 87

View Full Product Details