Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ANKRD29
Entrez Gene ID
101120373
Gene Name
ankyrin repeat domain 29
Gene Aliases
-
Chromosome Location
Chr.23: 37755523 - 37809166 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015103625.2 XP_014959111.1 6 - 7 633 64
XM_015103626.2 XP_014959112.1 6 - 7 610 64
XM_015103627.2 XP_014959113.1 5 - 6 516 64
XM_015103628.2 XP_014959114.1 5 - 6 498 64
XM_027960752.1 XP_027816553.1 5 - 6 591 64

View Full Product Details