Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRAP
Entrez Gene ID
105609272
Gene Name
melanocortin 2 receptor accessory protein
Gene Aliases
-
Chromosome Location
Chr.1: 131393281 - 131410677 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_012186386.3 XP_012041776.2 1 - 2 507 68
XM_027978352.1 XP_027834153.1 1 - 2 525 68
XM_027978362.1 XP_027834163.1 1 - 2 523 68

View Full Product Details