Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CSF3
Entrez Gene ID
443523
Gene Name
colony stimulating factor 3
Gene Aliases
CSF
Chromosome Location
Chr.11: 22848818 - 22851754 on Build Oar_rambouillet_v1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_027975456.1 XP_027831257.1 3 - 4 1234 91
GenBank mRNA L07939.1 - 3 - 4 361 91

View Full Product Details